3-Ethyl-4-hydroxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

28328-1G 28328-500MG 28328-5G 28328-25G
77468-130EA 140.08 USD
77468-130 77480-824 77491-176 77501-204
3-Ethyl-4-hydroxy-5-methylbenzaldehyde ≥97%
3-Ethyl-4-hydroxy-5-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD24673735
CAS Number: 42900-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR