Phenylmethyl 3-hydroxy-5-methylhexanoate ≥95%

Supplier: AOBChem USA

Warning

282387-1G 282387-500MG 282387-5G 282387-25G
77468-122EA 415.09 USD
77468-122 77480-816 77491-168 77501-196
Phenylmethyl 3-hydroxy-5-methylhexanoate ≥95%
Phenylmethyl 3-hydroxy-5-methylhexanoate

Precaución: For research use only

Formula: C₁₄H₂₀O₃
MW: 236.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34883913
CAS Number: 250221-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR