Phenylmethyl 3-hydroxy-3-methylbutanoate ≥95%

Supplier: AOBChem USA

Warning

282380-1G 282380-500MG 282380-5G 282380-25G
77468-118EA 237.93 USD
77468-118 77480-812 77491-164 77501-192
Phenylmethyl 3-hydroxy-3-methylbutanoate ≥95%
Phenylmethyl 3-hydroxy-3-methylbutanoate

Precaución: For research use only

Formula: C₁₂H₁₆O₃
MW: 208.25 g/mol
Storage Temperature: Ambient
CAS Number: 37526-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR