Isopropyl 3-cyclohexyl-3-hydroxypropanoate ≥97%

Supplier: AOBChem USA

Warning

281415-1G 281415-500MG 281415-5G 281415-25G
77468-114EA 415.09 USD
77468-114 77480-808 77491-160 77501-188
Isopropyl 3-cyclohexyl-3-hydroxypropanoate ≥97%
Isopropyl 3-cyclohexyl-3-hydroxypropanoate

Precaución: For research use only

Formula: C₁₂H₂₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34883590
CAS Number: 2969445-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR