(2,3-Difluoro-4-methylphenyl)triethylsilane ≥96%

Supplier: AOBChem USA

Warning

280851-1G 280851-500MG 280851-5G 280851-25G
77468-110EA 415.09 USD
77468-110 77480-804 77491-156 77501-186
(2,3-Difluoro-4-methylphenyl)triethylsilane ≥96%
(2,3-Difluoro-4-methylphenyl)triethylsilane

Precaución: For research use only

Formula: C₁₃H₂₀F₂Si
Storage Temperature: Ambient
MDL Number: MFCD34883271
CAS Number: 2969361-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR