1-(5-Bromo-2-fluoro-3-methylphenyl)pentan-1-ol ≥95%

Supplier: AOBChem USA

Warning

280847-1G 280847-500MG 280847-5G 280847-25G
77468-106EA 415.09 USD
77468-106 77480-800 77491-152 77501-182
1-(5-Bromo-2-fluoro-3-methylphenyl)pentan-1-ol ≥95%
1-(5-Bromo-2-fluoro-3-methylphenyl)pentan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO
MW: 275.1572 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34883268
CAS Number: 2969245-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR