(5-Bromo-2-fluoro-3-methylphenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

280843-1G 280843-500MG 280843-5G 280843-25G
77468-102EA 415.09 USD
77468-102 77480-796 77491-148 77501-178
(5-Bromo-2-fluoro-3-methylphenyl)(cyclopropyl)methanol ≥95%
(5-Bromo-2-fluoro-3-methylphenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₁H₁₂BrFO
Storage Temperature: Ambient
MDL Number: MFCD34883264
CAS Number: 2969452-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR