(5-Bromo-2-fluoro-3-methylphenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

280839-1G 280839-500MG 280839-5G 280839-25G
77468-098EA 415.09 USD
77468-098 77480-792 77491-144 77501-174
(5-Bromo-2-fluoro-3-methylphenyl)triethylsilane ≥95%
(5-Bromo-2-fluoro-3-methylphenyl)triethylsilane

Precaución: For research use only

Formula: C₁₃H₂₀BrFSi
MW: 303.2858 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34883260
CAS Number: 2969480-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR