Isopropyl 2-(1-hydroxycyclohexyl)acetate ≥97%

Supplier: AOBChem USA

Warning

280796-1G 280796-500MG 280796-5G 280796-25G
77468-094EA 415.09 USD
77468-094 77480-788 77491-140 77501-170
Isopropyl 2-(1-hydroxycyclohexyl)acetate ≥97%
Isopropyl 2-(1-hydroxycyclohexyl)acetate

Precaución: For research use only

Formula: C₁₁H₂₀O₃
Storage Temperature: Ambient
MDL Number: MFCD34883242
CAS Number: 54091-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR