2-(2-Fluorophenyl)tetrahydro-2H-pyran-2-ol ≥95%

Supplier: AOBChem USA

Warning

280793-1G 280793-500MG 280793-5G 280793-25G
77468-090EA 415.09 USD
77468-090 77480-784 77491-136 77501-166
2-(2-Fluorophenyl)tetrahydro-2H-pyran-2-ol ≥95%
2-(2-Fluorophenyl)tetrahydro-2H-pyran-2-ol

Precaución: For research use only

Formula: C₁₁H₁₃FO₂
MW: 196.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34883240
CAS Number: 2969400-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR