Ethyl(2-methoxy-4-methylphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

280399-1G 280399-500MG 280399-5G 280399-25G
77468-086EA 415.09 USD
77468-086 77480-780 77491-132 77501-162
Ethyl(2-methoxy-4-methylphenyl)sulfane ≥95%
Ethyl(2-methoxy-4-methylphenyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD34882919
CAS Number: 2969413-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR