3-(4-Methoxy-3,5-dimethylphenyl)-2,4-dimethylpentan-3-ol ≥97%

Supplier: AOBChem USA

Warning

279658-1G 279658-500MG 279658-5G 279658-25G
77468-082EA 415.09 USD
77468-082 77480-776 77491-128 77501-156
3-(4-Methoxy-3,5-dimethylphenyl)-2,4-dimethylpentan-3-ol ≥97%
3-(4-Methoxy-3,5-dimethylphenyl)-2,4-dimethylpentan-3-ol

Precaución: For research use only

Formula: C₁₆H₂₆O₂
MW: 250.3764 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34882612
CAS Number: 2969365-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR