Ethyl-4-(ethylthio)benzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(4-(Ethylthio)phenyl)-2-oxoacetate

279644-1G 279644-500MG 279644-5G 279644-25G
77468-078EA 278.1 USD
77468-078 77480-772 77491-124 77501-152
Ethyl-4-(ethylthio)benzoylformate ≥95%
Ethyl-4-(ethylthio)benzoylformate

Precaución: For research use only

Formula: C₁₂H₁₄O₃S
MW: 238.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801419
CAS Number: 300355-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR