2-Chloro-1-(4-(ethylthio)phenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

279637-1G 279637-500MG 279637-5G
77468-074EA 594.31 USD
77468-074 77480-768 77491-120
2-Chloro-1-(4-(ethylthio)phenyl)-2,2-difluoroethanone ≥97%
2-Chloro-1-(4-(ethylthio)phenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₉ClF₂OS
MW: 250.6926 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103835
CAS Number: 1823324-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR