1-(4-(ethylthio)phenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

279634-1G 279634-500MG 279634-5G
77468-070EA 594.31 USD
77468-070 77480-764 77491-116
1-(4-(ethylthio)phenyl)-2,2-difluoroethanone ≥97%
1-(4-(ethylthio)phenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₁₀F₂OS
MW: 216.2476 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103836
CAS Number: 1822670-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR