1-(4-(ethylthio)phenyl)-3-methylbutan-1-ol ≥97%

Supplier: AOBChem USA

Warning

279631-1G 279631-500MG 279631-5G 279631-25G
77468-066EA 312.09 USD
77468-066 77480-760 77491-112 77501-144
1-(4-(ethylthio)phenyl)-3-methylbutan-1-ol ≥97%
1-(4-(ethylthio)phenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀OS
MW: 224.3623 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34882597
CAS Number: 1695127-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR