1-(4-(ethylthio)phenyl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

279629-1G 279629-500MG 279629-5G 279629-25G
77468-062EA 312.09 USD
77468-062 77480-756 77491-108 77501-140
1-(4-(ethylthio)phenyl)-2-methylpropan-1-ol ≥97%
1-(4-(ethylthio)phenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₈OS
MW: 210.3357 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34882595
CAS Number: 1698132-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR