tert-Butyl (5-bromo-2-methoxyphenyl)carbamate ≥95%

Supplier: AOBChem USA

Warning

27939-1G 27939-500MG 27939-5G 27939-25G
77468-058EA 287.37 USD
77468-058 77480-752 77491-104 77501-136
tert-Butyl (5-bromo-2-methoxyphenyl)carbamate ≥95%
tert-Butyl (5-bromo-2-methoxyphenyl)carbamate

Precaución: For research use only

Formula: C₁₂H₁₆BrNO₃
MW: 302.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21170516
CAS Number: 262433-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR