(4-Chloro-5-fluoropyridin-3-yl)methano ≥95%

Supplier: AOBChem USA

Warning

27895-1G 27895-500MG 27895-5G 27895-25G
77468-054EA 368.74 USD
77468-054 77480-748 77491-100 77501-132
(4-Chloro-5-fluoropyridin-3-yl)methano ≥95%
(4-Chloro-5-fluoropyridin-3-yl)methano

Precaución: For research use only

Formula: C₆H₅ClFNO
MW: 161.561 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28142625
CAS Number: 1807166-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR