1-(4-Pentylphenyl)-1-propanone ≥96%

Supplier: AOBChem USA

Warning

278882-1G 278882-500MG 278882-5G 278882-25G
77468-046EA 175.1 USD
77468-046 77480-738 77491-090 77501-124
1-(4-Pentylphenyl)-1-propanone ≥96%
1-(4-Pentylphenyl)-1-propanone

Precaución: For research use only

Formula: C₁₄H₂₀O
MW: 204.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09373405
CAS Number: 17713-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR