2-(6-Bromo-3-chloro-2-fluorophenyl)pentan-2-ol ≥95%

Supplier: AOBChem USA

Warning

278800-1G 278800-500MG 278800-5G 278800-25G
77468-042EA 415.09 USD
77468-042 77480-736 77491-088 77501-122
2-(6-Bromo-3-chloro-2-fluorophenyl)pentan-2-ol ≥95%
2-(6-Bromo-3-chloro-2-fluorophenyl)pentan-2-ol

Precaución: For research use only

Formula: C₁₁H₁₃BrClFO
MW: 295.5757 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34882022
CAS Number: 2969245-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR