5-Chloro-4-methylthiophene carboxylic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
5-Chloro-4-methylthiophene-2-carboxylic acid
278768-1G
278768-500MG
278768-5G
278768-25G
77468-034EA
113.3
USD
77468-034
77480-728
77491-080
77501-114
5-Chloro-4-methylthiophene carboxylic acid ≥95%
5-Chloro-4-methylthiophene carboxylic acid
Precaución: For research use only
Formula:
C₆H₅ClO₂S Storage Temperature: Ambient |
MDL Number:
MFCD01830083 CAS Number: 74598-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...