2-Nitrothioanisole ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Methyl (2-nitrophenyl)sulphane, Methyl(2-nitrophenyl)sulphane

278729-1G 278729-500MG 278729-5G 278729-25G
77468-030EA 154.5 USD
77468-030 77480-724 77491-076 77501-110
2-Nitrothioanisole ≥95%
2-Nitrothioanisole

Precaución: For research use only

Formula: C₇H₇NO₂S
Storage Temperature: Ambient
MDL Number: MFCD03094691
CAS Number: 3058-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR