2,5-Dimethoxybenzophenone ≥95%

Supplier: AOBChem USA

Warning

278653-1G 278653-500MG 278653-5G
77468-026EA 576.8 USD
77468-026 77480-720 77491-072
2,5-Dimethoxybenzophenone ≥95%
2,5-Dimethoxybenzophenone

Precaución: For research use only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00769055
CAS Number: 4038-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR