3,6-Dichloro-2-(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

27855-1G 27855-500MG 27855-5G 27855-25G
77468-022EA 415.09 USD
77468-022 77480-716 77491-068 77501-106
3,6-Dichloro-2-(trifluoromethyl)benzaldehyde ≥95%
3,6-Dichloro-2-(trifluoromethyl)benzaldehyde

Precaución: For research use only

Formula: C₈H₃Cl₂F₃O
MW: 243.01 g/mol
Storage Temperature: Ambient
CAS Number: 2092790-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR