4-Cyclopropyl-α-methylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

278049-1G 278049-500MG 278049-5G
77468-018EA 508.82 USD
77468-018 77480-712 77491-064
4-Cyclopropyl-α-methylbenzenemethanol ≥96%
4-Cyclopropyl-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD23806193
CAS Number: 40641-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR