1-[2-(Trimethylsilyl)ethynyl]cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

277693-1G 277693-500MG 277693-5G
77468-014EA 443.93 USD
77468-014 77480-708 77491-060
1-[2-(Trimethylsilyl)ethynyl]cyclopentanol ≥95%
1-[2-(Trimethylsilyl)ethynyl]cyclopentanol

Precaución: For research use only

Formula: C₁₀H₁₈OSi
MW: 182.34 g/mol
Storage Temperature: Ambient
CAS Number: 71321-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR