1-[2-(Trimethylsilyl)ethynyl]cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

277691-1G 277691-500MG 277691-5G 277691-25G
77468-010EA 390.37 USD
77468-010 77480-704 77491-056 77501-104
1-[2-(Trimethylsilyl)ethynyl]cyclohexanol ≥95%
1-[2-(Trimethylsilyl)ethynyl]cyclohexanol

Precaución: For research use only

Formula: C₁₁H₂₀OSi
MW: 196.36 g/mol
Storage Temperature: Ambient
CAS Number: 17962-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR