2-Fluoro-α-[2-(trimethylsilyl)ethynyl]benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

277541-1G 277541-500MG 277541-5G 277541-25G
77468-006EA 390.37 USD
77468-006 77480-700 77491-052 77501-100
2-Fluoro-α-[2-(trimethylsilyl)ethynyl]benzenemethanol ≥95%
2-Fluoro-α-[2-(trimethylsilyl)ethynyl]benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₅FOSi
MW: 222.33 g/mol
Storage Temperature: Ambient
CAS Number: 1310487-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR