4-Chloro-α-[2-(trimethylsilyl)ethynyl]benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

277532-1G 277532-500MG 277532-5G
77468-002EA 447.02 USD
77468-002 77480-696 77491-048
4-Chloro-α-[2-(trimethylsilyl)ethynyl]benzenemethanol ≥95%
4-Chloro-α-[2-(trimethylsilyl)ethynyl]benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₅ClOSi
MW: 238.79 g/mol
Storage Temperature: Ambient
CAS Number: 1041849-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR