(6-Methoxy-4-methylpyridin-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

27749-1G 27749-500MG 27749-5G 27749-25G
77467-998EA 194.67 USD
77467-998 77480-692 77491-042 77501-092
(6-Methoxy-4-methylpyridin-3-yl)methanol ≥97%
(6-Methoxy-4-methylpyridin-3-yl)methanol

Precaución: For research use only

Formula: C₈H₁₁NO₂
MW: 153.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18905330
CAS Number: 1355190-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR