1-Bromo-2-isopropyl-4-benzyloxybenzene ≥97%

Supplier: AOBChem USA

Warning

27722-1G 27722-5G 27722-25G
77467-994EA 62.83 USD
77467-994 77491-038 77501-090
1-Bromo-2-isopropyl-4-benzyloxybenzene ≥97%
1-Bromo-2-isopropyl-4-benzyloxybenzene

Precaución: For research use only

Formula: C₁₆H₁₇BrO
Storage Temperature: Ambient
MDL Number: MFCD28142612
CAS Number: 211495-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR