(6-Bromo-3-ethoxy-2-fluorophenyl)(4-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

276431-1G 276431-500MG 276431-5G 276431-25G
77467-986EA 415.09 USD
77467-986 77480-682 77491-030 77501-082
(6-Bromo-3-ethoxy-2-fluorophenyl)(4-methoxyphenyl)methanol ≥95%
(6-Bromo-3-ethoxy-2-fluorophenyl)(4-methoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₆BrFO₃
MW: 355.1988 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34881474
CAS Number: 2969331-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR