2-Bromo-4,6-dichlorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

27541-1G 27541-500MG 27541-5G
77467-974EA 455.26 USD
77467-974 77480-670 77491-016
2-Bromo-4,6-dichlorobenzaldehyde ≥95%
2-Bromo-4,6-dichlorobenzaldehyde

Precaución: For research use only

Formula: C₇H₃BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD29088126
CAS Number: 1314031-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR