4-Methyl-5-(methylthio)nicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

27516-1G 27516-500MG 27516-5G
77467-970EA 767.35 USD
77467-970 77480-666 77491-012
4-Methyl-5-(methylthio)nicotinaldehyde ≥95%
4-Methyl-5-(methylthio)nicotinaldehyde

Precaución: For research use only

Formula: C₈H₉NOS
MW: 167.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26402595
CAS Number: 1289268-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR