2,5-Dimethoxy-α-methyl-α-(4-methylphenyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

274866-1G 274866-500MG 274866-5G 274866-25G
77467-966EA 415.09 USD
77467-966 77480-662 77491-008 77501-060
2,5-Dimethoxy-α-methyl-α-(4-methylphenyl)benzenemethanol ≥95%
2,5-Dimethoxy-α-methyl-α-(4-methylphenyl)benzenemethanol

Precaución: For research use only

Formula: C₁₇H₂₀O₃
Storage Temperature: Ambient
MDL Number: MFCD34881070
CAS Number: 1493016-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR