α-(4-Chlorophenyl)-3-propoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

274667-1G 274667-500MG 274667-5G 274667-25G
77467-962EA 415.09 USD
77467-962 77480-658 77491-004 77501-056
α-(4-Chlorophenyl)-3-propoxybenzenemethanol ≥95%
α-(4-Chlorophenyl)-3-propoxybenzenemethanol

Precaución: For research use only

Formula: C₁₆H₁₇ClO₂
MW: 276.76 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17140286
CAS Number: 1410118-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR