Benzenemethanol, 4-butyl-α-(3-chlorophenyl)- ≥95%

Supplier: AOBChem USA

Warning

274578-1G 274578-500MG 274578-5G 274578-25G
77467-958EA 415.09 USD
77467-958 77480-654 77491-000 77501-052
Benzenemethanol, 4-butyl-α-(3-chlorophenyl)- ≥95%
Benzenemethanol, 4-butyl-α-(3-chlorophenyl)-

Precaución: For research use only

Formula: C₁₇H₁₉ClO
MW: 274.79 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11096814
CAS Number: 944648-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR