3-Chloro-α-[4-(2-methylpropyl)phenyl]benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

274565-1G 274565-500MG 274565-5G 274565-25G
77467-954EA 415.09 USD
77467-954 77480-650 77490-996 77501-048
3-Chloro-α-[4-(2-methylpropyl)phenyl]benzenemethanol ≥95%
3-Chloro-α-[4-(2-methylpropyl)phenyl]benzenemethanol

Precaución: For research use only

Formula: C₁₇H₁₉ClO
MW: 274.79 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19146138
CAS Number: 1273934-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR