3-Chloro-α-[3-(1-methylethoxy)phenyl]benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

274522-1G 274522-500MG 274522-5G 274522-25G
77467-950EA 415.09 USD
77467-950 77480-646 77490-992 77501-044
3-Chloro-α-[3-(1-methylethoxy)phenyl]benzenemethanol ≥95%
3-Chloro-α-[3-(1-methylethoxy)phenyl]benzenemethanol

Precaución: For research use only

Formula: C₁₆H₁₇ClO₂
MW: 276.76 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19544842
CAS Number: 1472728-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR