α-[3-(Methylthio)phenyl]-2-pyridinemethanol ≥95%

Supplier: AOBChem USA

Warning

274229-1G 274229-500MG 274229-5G 274229-25G
77467-946EA 415.09 USD
77467-946 77480-642 77490-988 77501-040
α-[3-(Methylthio)phenyl]-2-pyridinemethanol ≥95%
α-[3-(Methylthio)phenyl]-2-pyridinemethanol

Precaución: For research use only

Formula: C₁₃H₁₃NOS
Storage Temperature: Ambient
CAS Number: 2400968-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR