α-[4-(Dimethylamino)phenyl]-2-pyridinemethanol ≥95%

Supplier: AOBChem USA

Warning

274204-1G 274204-500MG
77467-942EA 1466.72 USD
77467-942 77480-638
α-[4-(Dimethylamino)phenyl]-2-pyridinemethanol ≥95%
α-[4-(Dimethylamino)phenyl]-2-pyridinemethanol

Precaución: For research use only

Formula: C₁₄H₁₆N₂O
Storage Temperature: Ambient
CAS Number: 109520-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR