2-Ethoxy-1-isopropyl-4-methylbenzene ≥96%

Supplier: AOBChem USA

Warning

274156-1G 274156-500MG 274156-5G 274156-25G
77467-938EA 226.6 USD
77467-938 77480-634 77490-982 77501-036
2-Ethoxy-1-isopropyl-4-methylbenzene ≥96%
2-Ethoxy-1-isopropyl-4-methylbenzene

Precaución: For research use only

Formula: C₁₂H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD01707906
CAS Number: 4732-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR