Cyclopropyl(3-(methylthio)phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

273780-1G 273780-500MG 273780-5G
77467-934EA 522.21 USD
77467-934 77480-630 77490-978
Cyclopropyl(3-(methylthio)phenyl)methanol ≥95%
Cyclopropyl(3-(methylthio)phenyl)methanol

Precaución: For research use only

Formula: C₁₁H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD34149218
CAS Number: 2284036-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR