1-(2,4-dimethylphenyl)-3-methylbutan-1-ol ≥96%

Supplier: AOBChem USA

Warning

273777-1G 273777-500MG 273777-5G 273777-25G
77467-930EA 415.09 USD
77467-930 77480-626 77490-974 77501-030
1-(2,4-dimethylphenyl)-3-methylbutan-1-ol ≥96%
1-(2,4-dimethylphenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀O
MW: 192.2973 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787070
CAS Number: 858810-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR