2,4-Dichloro-5-hydroxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

27331-1G 27331-500MG 27331-5G 27331-25G
77467-926EA 229.69 USD
77467-926 77480-622 77490-970 77501-024
2,4-Dichloro-5-hydroxyphenylboronic acid ≥95%
2,4-Dichloro-5-hydroxyphenylboronic acid

Precaución: For research use only

Formula: C₆H₅BCl₂O₃
MW: 206.819 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015749
CAS Number: 1256346-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR