4-Bromo-3-fluoro-N,N-dimethylaniline ≥97%

Supplier: AOBChem USA

Warning

27231-1G 27231-500MG 27231-5G 27231-25G
77467-922EA 227.63 USD
77467-922 77480-618 77490-966 77501-018
4-Bromo-3-fluoro-N,N-dimethylaniline ≥97%
4-Bromo-3-fluoro-N,N-dimethylaniline

Precaución: For research use only

Formula: C₈H₉BrFN
Storage Temperature: Ambient
MDL Number: MFCD20920110
CAS Number: 1352218-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR