1-[1,1'-Biphenyl]-2-yl-1-propanone ≥95%

Supplier: AOBChem USA

Warning

271911-1G 271911-500MG 271911-5G
77467-918EA 487.19 USD
77467-918 77480-614 77490-960
1-[1,1'-Biphenyl]-2-yl-1-propanone ≥95%
1-[1,1'-Biphenyl]-2-yl-1-propanone

Precaución: For research use only

Formula: C₁₅H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD25977848
CAS Number: 92548-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR