5-Fluoro-4-methylpyridine-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

27143-1G 27143-500MG 27143-5G 27143-25G
77467-914EA 423.33 USD
77467-914 77480-610 77490-954 77501-010
5-Fluoro-4-methylpyridine-3-carboxylic acid ≥97%
5-Fluoro-4-methylpyridine-3-carboxylic acid

Precaución: For research use only

Formula: C₇H₆FNO₂
MW: 155.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257379
CAS Number: 1211584-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR