1-(2-Dibenzofuranyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

270594-1G 270594-500MG 270594-5G 270594-25G
77467-910EA 415.09 USD
77467-910 77480-606 77490-948 77501-004
1-(2-Dibenzofuranyl)-2,2,2-trifluoroethanone ≥95%
1-(2-Dibenzofuranyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₄H₇F₃O₂
MW: 264.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21338716
CAS Number: 129656-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR